missing translation for 'onlineSavingsMsg'
Learn More

Helios Italquartz Flat-Bottom Dishes with Spout, Transparent

Flat-bottom dishes with spouts constructed of transparent, durable, resistant, and chemically inert quartz glass.

Brand:  Helios Italquartz 61P00024

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 15808024

Quantity Price
1 166.00€ / Each
10 154.45€ / Each
Estimated Shipment: 18-10-2021

Add to basket


Description

Description

  • Transparent
  • Flat bottom
  • Includes spout
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications

Specifications

Flat
Round
100 mm
Evaporating Dish
Spout
20 mm
Transparent, includes spout
Quartz
90 cc
Videos
SDS
Documents
Certificates