Learn More
ACROBiosystems FITC-Labeled Human BCMA / TNFRSF17 Protein, Fc Tag DMF Filed
Brand: ACROBiosystems BCA-HF254-200UG
Additional Details : Weight : 0.00001kg
Description
Tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is also known as B-cell maturation protein (BCMA), CD antigen CD269, which is a member of the TNF-receptor superfamily. TNFRSF17 contains one TNFR-Cys repeat. TNFRSF17 is expressed in mature B-cells, but not in T-cells or monocytes. TNFRSF17 is receptor for TNFSF13B/BLyS/BAFF and TNFSF13/APRIL. TNFRSF17 promotes B-cell survival and plays a role in the regulation of humoral immunity. TNFRSF17 can activate NF-kappa-B and JNK.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 32.0 kDa. As a result of glycosylation, the protein migrates as 35-44 kDa under reducing (R) condition, and 60-75 kDa under non-reducing (NR) condition (SDS-PAGSpecifications
NP_001183.2 | |
200 μg | |
Met 1 - Ala 54 | |
TNFRSF17,CD269,BCM,BCMA | |
FITC | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |
32.0 kDa | |
FITC-Labeled Human BCMA, Fc Tag (Cat. No. BCA-HF254) is expressed from human HEK293 cells. It contains AA Met 1 - Ala 54 (Accession # Q02223-1). It is the FITC labeled form of Human BCMA, Fc Tag (Cat. No. BC7-H5254). | |
-20°C, avoid light | |
BCMA | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Fluorescent labeled protein |