Learn More
Fisherbrand™ Aluminum Weighing Dishes
Aluminum dishes for weighing and dispensing
Brand: Fisherbrand™
Additional Details : Weight : 5.60700kg
Description
Choose Fisherbrand™ Aluminum Weight Dishes for weighing and dispensing.
With Fluted sides
- Also can serve as evaporating dishes or dust covers Lightweight, yet sturdy Convenient finger-grip handle can be etched prior to weighing for identification
Specifications
Flat | |
Aluminum | |
Weighing Dish | |
20 mm | |
102 mm |
150 mL | |
Flanged Sides | |
For weighing and dispensing | |
96 mm | |
96 mm (Inner Base), 102 mm (Inner Top) |