Learn More
Traceable™ ISO 17034 (formerly ISO Guide 34) Color-Coded CRM pH Buffers (Certified Reference Material), Fisherbrand™
Performs accurate, reproducible pH calibrations with color-coded bottles. Fisherbrand™ Traceable, Color-Coded buffers are Certified Reference Material pH Buffers.Compatible with all makes of pH instruments and electrodes.
20.90€
Product Code | Brand | pH | Color | Chemical Name or Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH | Color | Chemical Name or Material | Price | Quantity & Availability | |||
15598693
|
Fisherbrand
|
4.005 | Red | Buffer Standards pH 4.005(CRM) |
20.90€
500mL |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||
15508703
|
Fisherbrand
|
7.000 | Yellow | Buffer Standards pH 7.000(CRM) |
20.90€
500mL |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||
15518703
|
Fisherbrand
|
10.012 | Blue | Buffer Standards pH 10.012(CRM) |
20.90€
500mL |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||
Description
Performs accurate, reproducible pH calibrations with color-coded bottles. Fisherbrand™ Traceable, Color-Coded buffers are Certified Reference Material pH Buffers. Compatible with all makes of pH instruments and electrodes.
- Certified Reference Material (CRM) solutions meets all test requirements for federal, state and local agencies
- 100% compatible with all makes of pH instruments and electrodes
- Long term buffer stability and buffer resistance to temperature changes during pH meter calibration
- Manufactured to IUPAC (International Union of Pure and Applied Chemistry) formulation
- Triple-layered proprietary packaging (aluminum, foil, PET and low-density polyethylene bag) prevents contamination by permeable gases
- Each 16 ounce bottle or six-pack single-use 100ml containers is supplied withindividual temperature chart, traceability information and Traceable™ Certificate