Learn More
Fisherbrand™ Traceable™ 5mL Vaccine Bottle Probe
Accessory for Radio-Signal Remote Thermometer
Brand: Fisherbrand™
Additional Details : Weight : 0.20000kg
Description
The Fisher Scientific™ Traceable™ 5mL Vaccine Bottle Probe monitors products subjected to door openings in freezers, incubators, and refrigerators
- Mimics vaccine temperature
- Fits in vaccine tray
- Filled with patented nontoxic glycol solution for temperature buffering
Specifications
Probe | |
Radio signal remote thermometer |