Learn More
Fisherbrand™ Pump Repair Kits
For use with vacuum pumps
Brand: Fisherbrand™
Additional Details : Weight : 1.00000kg
Description
For use with vacuum pumpsSpecifications
Model M8C and M12C vacuum pump | |
Gasket set, exhaust valves, valve springs, lip seals, O-ring, motor coupling | |
Pump Repair Kit |