Learn More
Macherey Nagel Filter Papers, MN 918 1/4, Folded
Medium fast, smooth, folded filter papers.
Brand: Macherey Nagel 561050
This item is not returnable.
View return policy
Description
- Smooth
- Medium fast filtration speed
- Suitable for the filtration of fruit juices, wines, and spirits
Specifications
| 120 g/m2 | |
| Folded | |
| MN 918 1/4 | |
| 0.34 mm |
| Smooth | |
| 9 sec. | |
| 100/Pk. | |
| 50 cm |