Learn More
Thermo Scientific™ SP6 RNA Polymerase, HC (≥100 U/µL)
Catalyze the 5'→3' synthesis of RNA on either single-stranded or double-stranded DNA with Thermo Scientific™ SP6 RNA polymerase.
75.00€ - 170.00€
Description
Highlights
- Incorporates modified nucleotides (e.g., aminoallyl-, biotin-, fluorescein-, digoxigenin-labeled nucleotides)