missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FcεR1α Mouse anti-Human, PE, Clone: AER-37 (also known as CRA-1, CRA1), BD Pharmingen™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566607
Additional Details : Weight : 0.00840kg
Description
FcεR1α Monoclonal antibody specifically detects FcεR1α in Human samples. It is validated for Flow Cytometry.Specifications
FcεR1α | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Human | |
Purified |
Flow Cytometry | |
AER-37 (also known as CRA-1, CRA1) | |
PE | |
0.1 mg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2b κ |