missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FBXW11 Recombinant Protein
335.00€ - 508.00€
Specifications
Accession Number | AAH26213 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 23291 |
Molecular Weight (g/mol) | 83.93 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16117132
|
Abnova™
H00023291-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16127132
|
Abnova™
H00023291-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH26213 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
83.93 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBXW11 | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
23291 | |
FBXW11 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEPDSVIEDKTIELMNTSVMEDQNEDESPKKNTLWQISNGTSSVIVSRKRPSEGNYQKEKDLCIKYFDQWSESDQVEFVEHLISRMCHYQHGHINSYLKPMLQRDFITALPEQGLDHIAENILSYLDARSLCAAELVCKEWQRVISEGMLWKKLIERMVRTDPLWKGLSERRGWDQYLFKNRPTDGPPNSFYRSLYPKIIQDIETIESNWRCGRHNLQRIQCRSENSKGVYCLQYDDEKIISGLRDNSIKIWDKTSLECLKVLTGHTGSVLCLQYDERVIVTGSSDSTVRVWDVNTGEVLNTLIHHNEAVLHLRFSNGLMVTCSKDRSIAVWDMASATDITLRRVLVGHRAAVNVVDFDDKYIVSASGDRTIKVWSTSTCEFVRTLNGHKRGIACLQYRDRLVVSGSSDNTIRLWDIECGACLRVLEGHEELVRCIRFDNKRIVSGAYDGKIKVWDLQAALDPRAPASTLCLRTLVEHSGRVFRLQFDEFQIISSSHDDTILIWDFLNVPPSAQNETRSPSRTYTYISR | |
BTRC2/BTRCP2/FBW1B/FBXW1B/Fbw11/Hos/KIAA0696 | |
FBXW11 | |
Wheat Germ (in vitro) | |
GST |