missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FASN (Human) Recombinant Protein
Human FASN full-length ORF ( AAH07909, 1 a.a. - 439 a.a.) recombinant protein with GST-tag at N-terminal.
335.00€ - 508.00€
Specifications
Accession Number | AAH07909 |
---|---|
Gene ID (Entrez) | 2194 |
Name | fatty acid synthase |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16124861
|
Abnova™
H00002194-P01.10UG |
10 μg |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16134861
|
Abnova™
H00002194-P01.25UG |
25 μg |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
- Sequence: MSTNDTIVSGTLPQRMASCLEVLDLFLNQPHMVLSSFVLAEKAAAYRDRDSQRDLVEAVAHILGIRDLAAVNLDSSLADLGLDSLMSVEVRQTLERELNLVLSVREVRQLTLRKLQELSSKADEASELACPTPKEDGLAQQQTQLNLRSLLVNPEGPTLMRLNSVQSSERPLFLVHPIEGSTTVFHSLASGLSIPTYGLQCTRAAPLDSIHSLAAYYIDCIRQVQPEGPYRVAGYSYGACVAFEMCSQLQAQQSPAPTHNSLFLFDGSPTYVLAYTQSYRAKLTPGCEAEAETEAICFFVQQFTDMEHNRVLEALLPLKGLEERVAAAVDLIIKSHQGLDRQELSFAARSFYYKLRAAEQYTPKAKYHGNVMLLRAKTGGAYGEDLGADYNLSQVCDGKVSVHVIEGDHRTLLEGSGLESIISIIHSSLAEPRVSVREG
Specifications
AAH07909 | |
fatty acid synthase | |
125% SDS-PAGE Stained with Coomassie Blue | |
FAS, MGC14367, MGC15706, OA-519, SDR27X1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
2194 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FASN | |
GST |