Learn More
Erlab FAS Molecular Filter
Product Code. 17126059 Shop All Erlab Products
Change view
Click to view available options
For Use With (Equipment):
Fume Hood
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17126059 Fume Hood Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17126059 Supplier Erlab Supplier No. H11112101
 Restricted Item
Estimated Shipment: 29-09-2026
Filter for your standalone filtration system Chemtrap H402.
  • Filter for your standalone filtration system Chemtrap H402
  • Type of filter: Molecular (Acid vapours)
  • Dimensions: 376 x 403 x 61 mm
  • Quantity/Set: 1
Specifications
For Use With (Equipment) Fume Hood
Product Type Carbon Filter