Learn More
Sigma Laborzentrifugen™ Falcon Adapters
Increase the flexibility of centrifugal applications with these adapters.
Brand: Sigma Laborzentrifugen™ 13060
Additional Details : Weight : 75.00000kg
Description
- Compatible with Sigma™ rotors and centrifuges
Specifications
15 mL Culture Tube, no. 15115 | |
Rotor 19776 | |
Gear |
2/Pk. | |
Adapts to 15mL culture, tube no. 15115 |