Learn More
Abnova™ FABP4 Recombinant Protein
Human FABP4 full-length ORF recombinant protein with GST-tag at N-terminal
335.00€ - 508.00€
Specifications
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
---|---|
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Molecular Weight (g/mol) | 40.26 |
pH Range | 8 |
Preparation Method | In vitro wheat germ expression system |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16164781
|
Abnova™
H00002167-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16174781
|
Abnova™
H00002167-P01.25UG |
25μg |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
FABP4 encodes the fatty acid binding protein found in adipocytes. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism.
- Molecular Weight: 40.26kDa
- Preparation Method: in vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 Fast Flow
- Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH 8.0 in the elution buffer
Sequence: MCDAFVGTWKLVSSENFDDYMKEVGVGFATRKVAGMAKPNMIISVNGDVITIKSESTFKNTEISFILGQEFDEVTADDRKVKSTITLDGGVLVHVQKWDGKSTTIKRKREDDKLVVECVMKGVTSTRVYERA
Best when used within three months from the date of receipt.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Antibody Production, ELISA, Protein Array, Western Blot | |
40.26 | |
In vitro wheat germ expression system | |
Wheat Germ (in vitro) | |
Recombinant |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Human | |
Solution |