missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol, 99.8%, anhydrous, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-7077273
Click to view available options
Quantity:
100 mL
1 L
107-21-1
C2H6O2
62.06
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
174
CHEBI:30742
ethane-1,2-diol
C(CO)O
107-21-1
C2H6O2
62.06
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
174
CHEBI:30742
ethane-1,2-diol
C(CO)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -13.0°C
Density 1.1130g/mL
Boiling Point 196.0°C to 198.0°C
Flash Point 116°C
Infrared Spectrum Authentic
Assay Percent Range 99.75% min. (GC)
Packaging AcroSeal™ Glass bottle
Linear Formula HOCH2CH2OH
Refractive Index 1.4300 to 1.4330
Quantity 100 mL
Merck Index 15,3852
Specific Gravity 1.113
Solubility Information Solubility in water: miscible. Other solubilities: miscible with acetic acid, acetone, glycerol,, pyridine, lower aliphatic alcohols,, aldehydes and ketones, slightly soluble in ether, practically insoluble in benzene and hydrocarbons
Viscosity 21 mPa.s (20°C)
Formula Weight 62.06
Percent Purity 99.8%
Chemical Name or Material Ethylene glycol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethylene glycol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'