Learn More
Ethylene-13C2-diamine 2HCl, CDN
Click to view available options
Quantity:
0.25 g
Unit Size:
0.25g
Specifications
Specifications
| Chemical Name or Material | Ethylenediamine-13C2 Dihydrochloride |
| CAS | 77435-60-0 |
| Molecular Formula | 2 Cl H . 13C2 H8 N2 |
| Quantity | 0.25 g |
| Synonym | 1,2-Ethanediamine-1,2-13C2, dihydrochloride (9 CI), 1,2-Ethanediamine-1,2-13C2 dihydrochloride, Ethylenediamine-13C2 dihydrochloride, Ethylene-13C2-diamine 2 HCl |
| InChI Formula | InChI=1 S/C2H8N2.2 ClH/c3-1-2-4;;/h1-4H2;2*1 H/i1+1,2+1;; |
| SMILES | Cl.Cl.N[13 CH2][13 CH2]N |
| IUPAC Name | (1,2-13C2)ethane-1,2-diamine;dihydrochloride |
| Molecular Weight (g/mol) | 135.0055 |
| Formula Weight | 134.0288 g/mol |
| Show More |