Learn More
Ethyl 5-(chloromethyl)-2-furancarboxylate, 95%, Thermo Scientific Chemicals
Catalog Number p-3778553
21.06 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
2528-00-9
C8H9ClO3
188.61
MFCD00173837
JBACYJRMCXLIQU-UHFFFAOYSA-N
ethyl 5-chloromethyl-2-furoate, ethyl 5-chloromethyl furan-2-carboxylate, ethyl 5-chloromethyl-2-furancarboxylate, 5-chloromethyl-2-furancarboxylic acid ethyl ester, 2-furancarboxylic acid, 5-chloromethyl-, ethyl ester, buttpark 9650-29, acmc-1cej4, ethyl 2-chloromethyl-5-furoate
263166
ethyl 5-(chloromethyl)furan-2-carboxylate
CCOC(=O)C1=CC=C(O1)CCl
2528-00-9
C8H9ClO3
188.61
MFCD00173837
JBACYJRMCXLIQU-UHFFFAOYSA-N
ethyl 5-chloromethyl-2-furoate, ethyl 5-chloromethyl furan-2-carboxylate, ethyl 5-chloromethyl-2-furancarboxylate, 5-chloromethyl-2-furancarboxylic acid ethyl ester, 2-furancarboxylic acid, 5-chloromethyl-, ethyl ester, buttpark 9650-29, acmc-1cej4, ethyl 2-chloromethyl-5-furoate
263166
ethyl 5-(chloromethyl)furan-2-carboxylate
CCOC(=O)C1=CC=C(O1)CCl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color Yellow
Density 1.2250g/mL
Boiling Point 115.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5100 to 1.5130
Quantity 1 g
Specific Gravity 1.225
Formula Weight 188.61
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl 5-(chloromethyl)-2-furancarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 5-(chloromethyl)-2-furancarboxylate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.