missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (S)-4-bromo-3-hydroxybutyrate, Thermo Scientific™
Click to view available options
Quantity:
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
CAS | 95537-36-3 |
Color | Colorless to Brown |
Molecular Formula | C6H11BrO3 |
MDL Number | MFCD00672875 |
Quantity | 1 g |
Synonym | s-ethyl 4-bromo-3-hydroxybutanoate, ethyl 3s-4-bromo-3-hydroxybutanoate, ethyl s-4-bromo-3-hydroxybutyrate, s-4-bromo-3-hydroxybutyric acid ethyl ether, ethyl s---4-bromo-3-hydroxybutyrate, ethyl-s---4-bromo-3-hydroxybutanoate, 4-bromo-2-ethyl-3-hydroxy-butanoate, ethyl 3s-4-bromo-3-hydroxybutyrate, s-3-hydroxy-4-bromobutyric acid ethyl ester, butanoic acid,4-bromo-3-hydroxy-,ethyl ester, 3r |
InChI Key | AIZRKZQHJNWBEI-YFKPBYRVSA-N |
SMILES | CCOC(=O)CC(CBr)O |
IUPAC Name | ethyl (3S)-4-bromo-3-hydroxybutanoate |
Molecular Weight (g/mol) | 211.055 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'