missing translation for 'onlineSavingsMsg'
Learn More

Ethyl pyrazolo[1,5-a]pyridine-3-carboxylate, 95%, Thermo Scientific™

Catalog Number p-3109978
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
16205-44-0
C10H10N2O2
190.20
MFCD06411542
IDHMFSBXNNIWCT-UHFFFAOYSA-N
ethyl pyrazolo 1,5-a pyridine-3-carboxylate, pyrazolo 1,5-a pyridine-3-carboxylic acid ethyl ester, ethylpyrazolo 1,5-a pyridine-3-carboxylate, ethyl h-pyrazolo 1,5-a pyridine-3-carboxylate, zlchem 54, acmc-1bsao, ethyl 3-azaindoline-3-carboxylate, ethyl 8-hydropyrazolo 1,5-a pyridine-3-carboxylate, 3-pyrazolo 1,5-a pyridinecarboxylic acid ethyl ester, 1qp
2795464
ethyl pyrazolo[1,5-a]pyridine-3-carboxylate
CCOC(=O)C1=C2C=CC=CN2N=C1
This item is not returnable. View return policy
16205-44-0
C10H10N2O2
190.20
MFCD06411542
IDHMFSBXNNIWCT-UHFFFAOYSA-N
ethyl pyrazolo 1,5-a pyridine-3-carboxylate, pyrazolo 1,5-a pyridine-3-carboxylic acid ethyl ester, ethylpyrazolo 1,5-a pyridine-3-carboxylate, ethyl h-pyrazolo 1,5-a pyridine-3-carboxylate, zlchem 54, acmc-1bsao, ethyl 3-azaindoline-3-carboxylate, ethyl 8-hydropyrazolo 1,5-a pyridine-3-carboxylate, 3-pyrazolo 1,5-a pyridinecarboxylic acid ethyl ester, 1qp
2795464
ethyl pyrazolo[1,5-a]pyridine-3-carboxylate
CCOC(=O)C1=C2C=CC=CN2N=C1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 190.2
Percent Purity 95%
Chemical Name or Material Ethyl pyrazolo[1, 5-a]pyridine-3-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'