missing translation for 'onlineSavingsMsg'
Learn More

Ethyl glycolate, 95%, Thermo Scientific Chemicals

Catalog Number p-4509543
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
623-50-7
C4H8O3
104.11
MFCD00021970
ZANNOFHADGWOLI-UHFFFAOYSA-N
ethyl glycolate, ethyl hydroxyacetate, acetic acid, hydroxy-, ethyl ester, glycolic acid, ethyl ester, ethyl glycollate, hydroxyacetic acid ethyl ester, glycolic acid ethyl ester, unii-c306e91wfk, acetic acid, 2-hydroxy-, ethyl ester, 2-hydroxyacetic acid ethyl ester
12184
ethyl 2-hydroxyacetate
CCOC(=O)CO
This item is not returnable. View return policy
623-50-7
C4H8O3
104.11
MFCD00021970
ZANNOFHADGWOLI-UHFFFAOYSA-N
ethyl glycolate, ethyl hydroxyacetate, acetic acid, hydroxy-, ethyl ester, glycolic acid, ethyl ester, ethyl glycollate, hydroxyacetic acid ethyl ester, glycolic acid ethyl ester, unii-c306e91wfk, acetic acid, 2-hydroxy-, ethyl ester, 2-hydroxyacetic acid ethyl ester
12184
ethyl 2-hydroxyacetate
CCOC(=O)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.0800g/mL
Boiling Point 158.0°C to 159.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula HOCH2COOCH2CH3
Refractive Index 1.4170 to 1.4200
Quantity 25 g
Specific Gravity 1.08
Formula Weight 104.11
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl glycolate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl glycolate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'