missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-hydroxyhexanoate, 97%, Thermo Scientific Chemicals

Catalog Number p-7081884
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
2305-25-1
C8H16O3
160.213
MFCD00036604
LYRIITRHDCNUHV-UHFFFAOYSA-N
hexanoic acid, 3-hydroxy-, ethyl ester, ethyl beta-hydroxycaproate, ethyl 3-hydroxy-hexanoate, ethyl hydroxy-3-hexanoate, 3-hydroxyhexanoic acid ethyl ester, fema no. 3545, acmc-20lakf, ethyl 3-hydroxycaproate, ethyl-3-hydrixyhexanoate, ethyl 3-hydroxy hexanoate
61293
CHEBI:23997
ethyl 3-hydroxyhexanoate
CCCC(CC(=O)OCC)O
This item is not returnable. View return policy
2305-25-1
C8H16O3
160.213
MFCD00036604
LYRIITRHDCNUHV-UHFFFAOYSA-N
hexanoic acid, 3-hydroxy-, ethyl ester, ethyl beta-hydroxycaproate, ethyl 3-hydroxy-hexanoate, ethyl hydroxy-3-hexanoate, 3-hydroxyhexanoic acid ethyl ester, fema no. 3545, acmc-20lakf, ethyl 3-hydroxycaproate, ethyl-3-hydrixyhexanoate, ethyl 3-hydroxy hexanoate
61293
CHEBI:23997
ethyl 3-hydroxyhexanoate
CCCC(CC(=O)OCC)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.974
Boiling Point 90°C to 92°C (14 mmHg)
Flash Point 94°C (202°F)
Linear Formula CH3CH2CH2CHOHCH2CO2CH2CH3
Refractive Index 1.4298
Quantity 25 g
Formula Weight 160.21
Percent Purity 97%
Chemical Name or Material Ethyl 3-hydroxyhexanoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'