Learn More
Saint-Gobain Ether-Based Polyurethane Tubing
Nontoxic, transparent, and sterilizable by ethylene oxide. Saint Gobain Performance Plastics™ Ether-based Polyurethane Tubing exhibits good elongation and recovery.
Brand:  Saint-Gobain 95869-06
Additional Details : Weight : 0.91000kg
Product Code. 11831930
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • Ether-grade polyurethane construction
  • Nontoxic
  • Transparent
  • Sterilizable by ethylene oxide.
  • Good elongation and recovery.
Specifications
Specifications
0.18 mm
8 mm
-40°C
Transparent
85°C
Polyurethane
30.48 m
1 Pack
330 psi