missing translation for 'onlineSavingsMsg'
Learn More

Ethanol-d{6}, anhydrous, 99+% (Isotopic), Thermo Scientific Chemicals

Catalog Number p-7049638
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1516-08-1
C2H6O
52.106
MFCD00051020
LFQSCWFLJHTTHZ-LIDOUZCJSA-N
ethanol-d6, 2h6 ethanol, hexadeuteroethanol, 2 h? ethan 2 h ol, ethyl-d5 alcohol-d, ethyl alcohol-d6, ethanol d, ethanol-d6 9ci
102138
1,1,1,2,2-pentadeuterio-2-deuteriooxyethane
CCO
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1516-08-1
C2H6O
52.106
MFCD00051020
LFQSCWFLJHTTHZ-LIDOUZCJSA-N
ethanol-d6, 2h6 ethanol, hexadeuteroethanol, 2 h? ethan 2 h ol, ethyl-d5 alcohol-d, ethyl alcohol-d6, ethanol d, ethanol-d6 9ci
102138
1,1,1,2,2-pentadeuterio-2-deuteriooxyethane
CCO

NMR Solvent

Ethanol-d_6, is employed as an NMR solvent. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Color Colorless
Density 0.91 g/mL
Odor Alcohol-like
Sensitivity Hygroscopic
Quantity 1 g
Boiling Point 79°C
Linear Formula CD3CD2OD
UN Number UN1170
Solubility Information Soluble in water.
Formula Weight 52.11
Percent Purity ≥99%
Chemical Name or Material Ethanol-d6, Isotopic, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'