missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, Absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC), Honeywell™

Brand:  Honeywell Research Chemicals 24194-30KG

Additional Details : CAS : 64-17-5 Weight : 30.00000kg Transport : UN number : 1170 Chem class : 3 Pack group : II

Quantity 30kg

 View more versions of this product

Product Code. 15622540

  • 639.55€ / 30kg


please call +351214253350
Add to basket

Specifications

Chemical Identifiers

64-17-5
46.069
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
702
ethanol
C2H6O
MFCD00003568
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol
CHEBI:16236
CCO

Specifications

Ethanol
64-17-5
CH3CH2OH
1170
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
702
46.07g/mol
0.789g/mL at 25°C
683°F
1.59 (vs air)
30kg
Absolute
C2H6O
MFCD00003568
1718733
Completely souble in water
CCO
46.069
CHEBI:16236
≥98% (GC)
19% (60°F)
44.6mmHg (20°C)
14°C (57.2°F)
Videos
Documents