Learn More
Abnova™ ETF1 (Human) Recombinant Protein
Human ETF1 partial ORF ( NP_004721.1, 338 a.a. - 437 a.a.) recombinant protein with GST-tag at N-terminal.
335.00€ - 508.00€
Specifications
Accession Number | NP_004721.1 |
---|---|
Gene ID (Entrez) | 2107 |
Name | eukaryotic translation termination factor 1 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16117814
|
Abnova™
H00002107-Q01.10UG |
10 μg |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16127814
|
Abnova™
H00002107-Q01.25UG |
25 μg |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
- Sequence: TEEEKILYLTPEQEKDKSHFTDKETGQEHELIESMPLLEWFANNYKKFGATLEIVTDKSQEGSQFVKGFGGIGGILRYRVDFQGMEYQGGDDEFFDLDDY
Specifications
NP_004721.1 | |
eukaryotic translation termination factor 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
D5S1995, ERF, ERF1, MGC111066, RF1, SUP45L1, TB3-1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
2107 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ETF1 | |
GST |