Learn More
Thermo Scientific Chemicals Esmolol hydrochloride
Product Code. 15862484 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 15862484
Brand: Thermo Scientific Chemicals 462460050
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 81161-17-3
Molecular Formula C16H26ClNO4
Molecular Weight (g/mol) 331.84
InChI Key GEKNCWBANDDJJL-UHFFFAOYNA-N
IUPAC Name methyl 3-(4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)propanoate hydrochloride
SMILES Cl.COC(=O)CCC1=CC=C(OCC(O)CNC(C)C)C=C1
Specifications
Melting Point 86.0°C to 92.0°C
Quantity 5 g
Formula Weight 331.83
Chemical Name or Material Esmolol hydrochloride
compliance-icons
Product Identifier
  • Esmolol hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Esmolol hydrochloride