missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Eriochrome™Black T, ACS reagent, indicator grade
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Clarity | (0.1%, water) Passes Test |
Solubility Information | Solubility in water: slightly soluble. |
IUPAC Name | (4Z)-4-[(1-hydroxynaphthalen-2-yl)hydrazinylidene]-7-nitro-3-oxonaphthalene-1-sulfonic acid;sodium |
Formula Weight | 461.38 |
Grade | ACS Reagent |
Packaging | Glass bottle |
Suitability | Suitability as complexometric indicator: Passes Test |
Color | Black/Brown |
Quantity | 250 g |
Physical Form | Powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'