Learn More
ERI2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557251
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TVPISDSDLE ISFNSGERLM VLKELEMSSH ENFGDIEETP QKSETSKSIV YKSPHTTIYN VKEAKDPGSD ISAFKLPEHK SSTFNRVNAN MSHPLVLGKH PLLSGG Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 64%.
ERI2 Polyclonal antibody specifically detects ERI2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
ERI2 | |
Polyclonal | |
Unconjugated | |
ERI2 | |
enhanced RNAi three prime mRNA exonuclease homolog 2; ERI1 exoribonuclease 2; ERI1 exoribonuclease family member 2; ERI2; EXOD1; exonuclease domain containing 1; exonuclease domain-containing protein 1; exoribonuclease 2; KIAA1504; ZGRF5; zinc finger, GRF-type containing 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
112479 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A8K979 | |
ERI2 | |
Recombinant protein corresponding to Human ERI2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only