Learn More
Eppendorf™ epTIPS™ Dual Filter Tips
Use these pipette tips for full sample recovery and quick pipetting. epTIPS™ Dual Filter Tips offer a two-phase filter for contamination protection. The two filter layers made of flexible, hydrophobic material, fit perfectly in the tip cone and retain near 100% of all aerosols and biomolecules.
POA
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
Clean Claims | PCR clean |
Compatibility | Universal Fit Pipettor |
Tip Style | Universal Fit |
Description
Use these pipette tips for full sample recovery and quick pipetting. epTIPS™ Dual Filter Tips offer a two-phase filter for contamination protection. The two filter layers made of flexible, hydrophobic material, fit perfectly in the tip cone and retain near 100% of all aerosols and biomolecules.
Tips consist of a unique two phase filter protection barrier made from hydrophobic polyethylene:- The upper blue layer provides very effective protection against small aerosols and biomolecules (DNA)
- The lower layer acts as pre-filter and efficiently protects against larger aerosols, drops, splashes and larger amounts of liquids
- The combination of both layers leads to a unique efficiency against aerosol contaminants
- All ep Dualfilter T.I.P.S.™ are PCR* clean (free of human DNA, DNase, RNase, PCR* inhibitors) and sterile (to USP, DAB, Ph. Eur. standards) to satisfy the particular requirements of the molecular biology laboratory
Specifications
Filtered | |
PCR clean | |
Universal Fit |
Sterile | |
Universal Fit Pipettor | |
Universal Fit Pipettor |