Learn More
Eppendorf™ epTIPS™ LoRetention Tips
Unique material property provides maximum sample recovery, improved reproducibility and higher sensitivity for detergent containing liquids
Brand:  Eppendorf™ 0030072286
Additional Details : Weight : 0.76500kg
 View more versions of this product
Product Code. 11873462
  • 61.58€ / Pack of 480
Estimated Shipment
10-02-2021
Add to basket
Description
Description
  • Homogenous surface achieved through treatment at the molecular level — Pearl Effect Technology
  • Tips are uncoated and free from additives
  • Ultra-homogenous surface for maximum reproducibility
  • Ultra-hydrophobic surface for minimum loss of sample
  • Air bubbles and foaming are significantly reduced during pipetting
  • Autoclavable, high chemical resistance
  • Five-year shelf life
Specifications
Specifications
50 to 1000 μL
Autoclavable
Unique material property provides maximum sample recovery, improved reproducibility and higher sensitivity for detergent containing liquids
Blue
7.1cm
Low Retention