Learn More
Eppendorf™ ep Dualfilter T.I.P.S.™ Filter Tips
Protects pipette and sample against unintentional and unnoticed contamination by aerosols, drops and splashes. Eppendorf™ ep Dualfilter T.I.P.S.™ Filter Tips has two-phase filter for contamination protection.
Brand:  Eppendorf™ 0030077784
Additional Details : Weight : 1.20150kg
 View more versions of this product
Product Code. 15625357
  • 193.60€ / Pack of 960
Estimated Shipment
05-08-2019
Add to basket
Description
Description
  • Two filter layers
  • Provides dual protection against aerosols and biomolecules to guard both pipette and sample
  • Free of PCR inhibiting additives
  • Provide reliable downstream results
  • Provides high purity and quality
Applications
  • DNA applications (e.g. PCR)
  • RNA applications (e.g. Gen expression analysis)
  • Protein applications (e.g. Antibody Research)
  • Cell Culture applications (e.g. Media)
  • Applications with radio actives
  • All applications which use aerosol binding liquids
Specifications
Specifications
Rack
Yellow
DNA, RNA and Protein applications
2 to 200μL
EN 1822