Learn More
Enzymaster Enzyme Screening Kit
Product Code. 17894621
Click to view available options
For Use With (Application):
Enzyme screening of aldolases
Enzyme screening of iminereductases
Enzyme screening of ketoreductases
Enzyme screening of transaminases
Kit Contents:
16 vials with 0.5 g enzyme (CFE)
20 vials with 0.5 g enzyme (CFE)
21 vials with 0.5 g enzyme (CFE)
Unit Size:
Each
Product Code. 17894621
Brand: Enzymaster 200400
Only null left
Add to basket
Add to basket
The Enzyme Screening Kit contains a wide range of wild type and engineered enzymes with a broad substrate scope for your screening!
  • Ketoreductase Kit
    • Enzyme kit containing 20 ketoreductases (E.C. 1.1.1.) for a quick screening
    • Applicable for ketone reduction and alcohol oxidation
    • Diverse range of substrates possible, both (S)- and (R)-selective enzymes included
    • 0.5 g per enzyme
    • Formulated as lyophilized cell-free extract (CFE)
  • IRED Kit
    • Enzyme kit containing 16 iminereductases (E.C. 1.5.1.) for a quick screening
    • Applicable for imine reduction and reductive amination
    • Diverse range of substrates possible, both (S)- and (R)-selective enzymes included
    • 0.5 g per enzyme
    • Formulated as lyophilized cell-free extract (CFE)
  • Alolase Kit
    • Enzyme kit containing 20 aldolases (E.C. 4.1.2.) for a quick screening
    • Contains threonine, phenylserine and serine aldolases
    • Applicable for reversible aldol reaction of glycine and an aldehyde to β-hydroxy-α-l-amino acid
    • Diverse range of substrates possible, high selectivity
    • 0.5 g per enzyme
    • Formulated as lyophilized cell-free extract (CFE)
  • Transaminase Kit
    • Enzyme kit containing 21 transaminases (E.C. 2.6.1.) for a quick screening
    • Applicable for reversible transamination of an amino group from an amino donor to a keto amino acceptor
    • Diverse range of substrates possible, both (S)- and (R)-selective enzymes included
    • 0.5 g enzyme powder per vial
    • Formulated as lyophilized cell-free extract (CFE)
Specifications
Product Type Enzyme Screening Kit
Kit Contents 21 vials with 0.5 g enzyme (CFE)
For Use With (Application) Enzyme screening of transaminases