Learn More
Ark-Plas Products™ Enlarging Check Valve
Created from Kynar. Ark-Plas Products™ Enlarging Check Valve is excellent for prevention of backflow of fluid. The valve allows for an increase in pressure of the fluid.
Brand: Ark-Plas Products™ HDX20-DF0
Additional Details : Weight : 0.03960kg
Description
- Created from Kynar polypropylene
- Ideal for preventing backflow of fluid
Specifications
Natural | |
Kynar | |
3/16 x 1/4 in. |