Learn More
ACROBiosystems ELISA Assay Kit for Anti-CTLA-4 h-mAb in Human Serum
For quantification of monoclonal anti-CTLA-4 antibodies in human serum.
Brand:  ACROBiosystems ECH-V1-480TESTS
Additional Details : Weight : 1.50000kg
 View more versions of this product
Product Code. 15826607
  • 1528.32€ / 1 set
Estimated Shipment: 25-11-2021
Add to basket
Description
Description
How a therapeutic antibody is metabolized in the body is pertinently relevant to its efficacy. Therefore, pharmacokinetics study is an important part of the drug development. CTLA-4 is probably the most targeted molecule in today’s pharmaceutical industry, thanking to the clinical success of Yervoy®. Many investigational anti-CTLA-4 mAbs are being developed. There is a growing need for a standard assay that can be used to facilitate the study of their pharmacokinetics.
Specifications
Specifications
Pk. Assay Kit
-20°C, aviod light
RT
The enclosed ELISA assay kit for anti-CTLA-4 h-mAb in human serum is based on an enzyme immunoassay (ELISA) between recombinant CTLA-4 protein (ECD) and a biotinylated anti-CTLA-4 monoclonal antibody. The method employs the principle of competitive ELISA
480 Tests