missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems ELISA Assay Kit for Anti-CTLA-4 h-mAb in Human Serum
For quantification of monoclonal anti-CTLA-4 antibodies in human serum.
561.77€
Specifications
Product Type | Pk. Assay Kit |
---|---|
Kit Contents | The enclosed ELISA assay kit for anti-CTLA-4 h-mAb in human serum is based on an enzyme immunoassay (ELISA) between recombinant CTLA-4 protein (ECD) and a biotinylated anti-CTLA-4 monoclonal antibody. The method employs the principle of competitive ELISA |
Shipping Condition | RT |
Storage Requirements | -20°C, aviod light |
Description
How a therapeutic antibody is metabolized in the body is pertinently relevant to its efficacy. Therefore, pharmacokinetics study is an important part of the drug development. CTLA-4 is probably the most targeted molecule in today’s pharmaceutical industry, thanking to the clinical success of Yervoy®. Many investigational anti-CTLA-4 mAbs are being developed. There is a growing need for a standard assay that can be used to facilitate the study of their pharmacokinetics.Specifications
Pk. Assay Kit | |
The enclosed ELISA assay kit for anti-CTLA-4 h-mAb in human serum is based on an enzyme immunoassay (ELISA) between recombinant CTLA-4 protein (ECD) and a biotinylated anti-CTLA-4 monoclonal antibody. The method employs the principle of competitive ELISA | |
RT | |
-20°C, aviod light |