missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Electronic Crimping Tools, Handheld, Battery

Reduces hand strain compared to manual crimper operation.

Brand:  Fisherbrand™

Additional Details : Weight : 2.00000kg Transport : UN number : 3481 Chem class : 9

 View more versions of this product

Product Code. 15502340

  • 975.00€ / Each


Estimated Shipment date
from Supplier 18-02-2019
Add to basket

Description

Description

  • Electronic Crimper
  • 110 - 240V
  • Electronic
  • Steel/plastic
  • Crimper
  • Adjustable
  • For Aluminum/Steel caps
Specifications

Specifications

No
20mm
Electronic
20mm Electronic Crimper
Aluminum Cap
Videos