missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDNRA/Endothelin R Type A Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99333C
This item is not returnable.
View return policy
Description
EDNRA/Endothelin R Type A Polyclonal specifically detects EDNRA/Endothelin R Type A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EDNRA/Endothelin R Type A | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
endothelin receptor type A, endothelin-1 receptor, endothelin-1-specific receptor, ET1-specific type, ETAR, ETRA, G protein-coupled receptor, hET-AR | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human EDNRA/Endothelin R Type A. | |
0.1 mL | |
GPCR, Neuroscience, Signal Transduction | |
1909 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |