Learn More
S.C.A.T™ Exhaust filter with Change Indicator for SCAT SafetyWasteCaps
The S.C.A.T™ exhaust filter adsorbs solvent vapors when collecting liquid waste. The carbon is steam activated and developed to capture vapors of organic solvents. Activate the change indicator by pushing the button, and stay informed about the elapsed service time.
238.83€ - 257.09€
Specifications
For Use With (Equipment) SCAT SafetyWasteCaps
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15179694 View Documents S.C.A.T™
190336
3 Pieces 257.09€
Pack of 3 View alternative products
Save Up to 
»
In Stock 19
Add to basket
15317872 View Documents S.C.A.T™
190335
4 Pieces 238.83€
Pack of 4 View alternative products
Save Up to 
»
Estimated Shipment
28-10-2019
Add to basket
Description
Description
  • Core element of S.C.A.T™ SafetyWasteCaps
  • Prevents inhalation of harmful solvent vapors
  • Steam activated carbon adsorbs 99% of volatile substances
  • Optimal ventilation slots ensure constant flow of filtered exhaust air
  • Easy screwing on S.C.A.T™ SafetyWasteCaps
  • Includes change indicator and splash protection
  • Available in 3 Sizes S, M, L
  • To be changed every 3 or 6 months 
  • Lab safety, optimal protection during disposal of solvents
  • Safety disposal of HPLC waste
  • Technical protective measures
  • Avoid skin contact with organic solvents
  • Create a comfortable working climate
Specifications
Specifications
SCAT SafetyWasteCaps