Learn More
Terumo™ Eccentric Luer-Slip Tip Three-Part Syringe
Terumo™ Eccentric Luer-Slip Tip Three-Part Syringes are easy to use with their smooth plunger action and have clear barrels to allow quick recognition of air bubbles for more accurate dosing. With easy to read markings, you can be confident you are delivering the intended dose every time.
12.51€
Specifications
Material Polypropylene
Disposable Yes
Sterility Sterile
View More Specs
Products 1
Product Code Brand Volume (Metric) Product Type Quantity Price Quantity & Availability  
Product Code Brand Volume (Metric) Product Type Quantity Price Quantity & Availability  
11793313
View Documents Promotion Details
Terumo™
1SS02LE1
2.5 mL Centric Luer-lock 3-part Syringe 100/Pk.
12.51€
Pack of 100
View alternative products
Save Up to 
»
In Stock
Description
Use these syringes for the aspiration of fluids or sampling blood or other body fluids. Terumo™ Eccentric Luer-Tip Three-Part Syringe feature a unique double beveled design for greater patient comfort.
  • Silicone coated barrel for a smooth plunger action
  • Clear barrel to allow quick recognition of air bubbles
  • Easy to read markings
  • Latex free
  • Sterile and packed in individually wrapped tamper peel packs
  • Syringes are available with luer lock, luer slip, catheter and eccentric tips
  • Available in a wide range of sizes