Learn More
Macherey Nagel EC HPLC column EC 100/4NUCLEODUR SPHINX RP, 3 µm length: 100 mm,
Brand:  Macherey Nagel 760812.40
EC HPLC column EC 100/4NUCLEODUR SPHINX RP, 3 µm length: 100 mm,
Product Code. 12743501
  • 826.00€ / Each
Estimated Shipment: 21-01-2025