Learn More
Abeba™ Dynamic 7362 Shoes
Special respiration absorbing lining allows the foot to breathe. Includes TPU sole with high slip resistance.
Brand:  Abeba™ 7362/39
Additional Details : Weight : 0.99000kg
 View more versions of this product
Product Code. 15622808
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Microfiber
  • Gray/Blue
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3574)
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Microfiber
Slip-resistant
7362
Gray, Blue
39
Polyurethane