Learn More
DWK Life Sciences DURAN™ Measuring Cylinder with UKAS Certificate, Class A
The DURAN™ Class A measuring cylinders with UKAS certificate are calibrated under defined conditions in accordance with ISO 1042, DIN 12664 and ISO 17025 standards.
Brand: DWK Life Sciences 213975406
202.35 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.63000kg
Description
Our DURAN™ volumetric measuring instruments already meet the highest accuracy standard. The DURAN™ range has now been expanded to include the product range of volumetric flasks and measuring cylinders with UKAS individual certificates. This new product line is tested and calibrated according to the requirements of the United Kingdom Accreditation Services (UKAS) and complies with ISO 17025, a standard for worldwide comparability of test and calibration results.
- The volume content tolerances conform to accuracy Class A
- High precision due to better visibility of the printed ring mark
- The blue printed image is highly chemically resistant
- Easy-to-read scale with graduation marks to estimate the content
- Thanks to the exact processing and precisely calibrated scales, they permit the highly accurate determination and measurements of volume
- Maximum minimization of unwanted influences
Specifications
5 mL | |
65 mm | |
Measuring liquid amounts; Diluting solutions; Mixing several components; Volumetric measurement | |
Yes |
Borosilicate Glass (3.3) | |
1000 mL | |
1/Pk. |