Learn More
Fisherbrand™ Dummy Plate
Accessory for use with Vertical Gel Electrophoresis System.
12.00€ - 15.45€
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
11854502
|
Fisherbrand™
|
Dummy Plate, 200 x 200 mm |
15.45€
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
11877623
|
Fisherbrand™
|
Dummy Plate, 100 x 100 mm |
12.00€
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
Description
- Accessory for use with Vertical Gel Electrophoresis System
Specifications
Vertical Gel Electrophoresis System |