Learn More
Dräger™ Short-Term Detector Tubes: Vinyl Chloride
Detector tubes
212.00€ - 215.00€
Specifications
Components Measured Vinyl Chloride
Quantity 10
View More Specs
Products 2
Product Code Brand Components Measured Tube Type Measurement Range Humidity Range Price Quantity & Availability  
Product Code Brand Components Measured Tube Type Measurement Range Humidity Range Price Quantity & Availability  
13408806
missing translation for 'viewDocuments' Promotion Details
Dräger™
CH19601
Vinyl Chloride Vinyl chloride 1/a 100 to 3000 ppm Less than 30 mg/L
215.00€
Each
Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 10-05-2024
to see stock.
Zum Warenkorb hinzufügen
10754253
missing translation for 'viewDocuments' Promotion Details
Dräger™
8101721
Vinyl Chloride Vinyl chloride 0.5/b 5 to 30 20mg/L
212.00€
Each
Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 15-05-2024
to see stock.
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
 Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Bestellinformation
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Spezifikation
Spezifikation
Vinyl Chloride
10
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate