missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DPNB-ABT 594
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
DPNB-ABT 594 is a nitrobenzyl-caged ABT 594 (Cat. No. 6576), a selective α4β2 nAChR agonist. One-photon uncaging evokes large inward currents and Ca2+ transients on cell bodies and dendrites of medial habenular neurons in mouse brain slices. Two-photon uncaging induces fast nAChR-mediated currents. Photolyzed with high quantum yield of 0.20. Effective photolysis occurs using one- or two-photon excitation; one-photon uncaging requires illumination at 410 nm for 1.5-3 ms; two-photon uncaging requires illumination with appropriate two photon pulse laser at 710 nm for ∽2 ms.
Specifications
Specifications
| Chemical Name or Material | 5-(((2 R)-1-(1-(4,5-Bis(2-(2-(2-methoxyethoxy)ethoxy)ethoxy)-2-nitrophenyl)ethyl)azetidin-2-yl)methoxy)-2-chloropyridine |
| Quantity | 5 mg |
| Target | Nicotinic (a4b2) Receptor Agonists |
| Molecular Formula | C31H46ClN3O11 |
| Formula Weight | 672.16 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction