Learn More
LMS™ Double Glazed Door for Cooled Incubator
LMS™ Cooled Incubators come with a wide range of factory-fitted optional accessories and other extras to cater for individual requirements and even more operational flexibility.
Brand: LMS™ DG14
Additional Details : Weight : 0.80000kg
Description
- Double glazed glass door
- Optional factory-fitted alternative for the LMS™ Model 200 or 210 cooled incubators
- Provides total visibility of incubator contents
- Incubator temperature range is adjusted from 0°C to +50°C if these glass doors are selected
Order Info
ForUseWithApplication
Specifications
Exterior glass door | |
LMS™ Model 210 Cooled Incubators 135L | |
Double Glazed Dorr | |
2 Years |