missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DLX2 (Human) Recombinant Protein
Human DLX2 partial ORF (NP_004396.1, 1 a.a. - 110 a.a.) recombinant protein with GST tag at N-terminal.
Brand: Abnova™ H00001746-Q02.10ug
Additional Details : Weight : 0.02000kg
Description
- Sequence: MTGVFDSLVADMHSTQIAASSTYHQHQQPPSGGGAGPGGNSSSSSSLHKPQESPTLPVSTATDSSYYTNQQHPAGGGGGGGSPYAHMGSYQYQASGLNNVPYSAKSSYDL
Specifications
NP_004396.1 | |
distal-less homeobox 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DLX2 | |
GST |
1746 | |
Wheat germ expression system | |
10 μg | |
TES-1, TES1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |