missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DLX2 (Human) Recombinant Protein
Human DLX2 partial ORF (NP_004396.1, 1 a.a. - 110 a.a.) recombinant protein with GST tag at N-terminal.
335.00€ - 508.00€
Specifications
Accession Number | NP_004396.1 |
---|---|
Gene ID (Entrez) | 1746 |
Name | distal-less homeobox 2 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16033905
|
Abnova™
H00001746-Q02.10UG |
10 μg |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16043905
|
Abnova™
H00001746-Q02.25UG |
25 μg |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
- Sequence: MTGVFDSLVADMHSTQIAASSTYHQHQQPPSGGGAGPGGNSSSSSSLHKPQESPTLPVSTATDSSYYTNQQHPAGGGGGGGSPYAHMGSYQYQASGLNNVPYSAKSSYDL
Specifications
NP_004396.1 | |
distal-less homeobox 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
TES-1, TES1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
1746 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DLX2 | |
GST |