missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Tropic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
It is a laboratory reagent used in the chemical synthesis of atropine and hyoscyamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a laboratory reagent used in the chemical synthesis of atropine and hyoscyamine.
Solubility
Soluble in water (20 g/L) at 20°C, ethanol, ether, alcohol, and methanol (0.1 g/ml).
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
It is a laboratory reagent used in the chemical synthesis of atropine and hyoscyamine.
Solubility
Soluble in water (20 g/L) at 20°C, ethanol, ether, alcohol, and methanol (0.1 g/ml).
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Specifications
Melting Point | 117°C to 119°C |
Quantity | 10 g |
Beilstein | 2209199 |
Merck Index | 14,9779 |
Solubility Information | Soluble in water (20g/L) at 20°C,ethanol,ether,alcohol,and methanol (0.1g/ml). |
Formula Weight | 166.18 |
Percent Purity | 98% |
Chemical Name or Material | DL-Tropic acid |
Safety and Handling
Product Identifier
- DL-Tropic acid
- Warning
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'