missing translation for 'onlineSavingsMsg'
Learn More

DL-10-Camphorsulfonic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-82181
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
5872-08-2
C10H16O4S
232.29
MFCD00074827
MIOPJNTWMNEORI-UHFFFAOYNA-N
reychler's acid, camphorsulfonic acid, d-camphorsulfonic acid, camphersulfosaeure, --10-camphorsulfonic acid, 2-oxobornane-10-sulphonic acid, d-10-camphorsulfonic acid, 7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, l-camphor-10-sulfonic acid, +-10-camphorsulfonic acid
18462
CHEBI:55379
(7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl)methanesulfonic acid
CC1(C2CCC1(C(=O)C2)CS(=O)(=O)O)C
This item is not returnable. View return policy
5872-08-2
C10H16O4S
232.29
MFCD00074827
MIOPJNTWMNEORI-UHFFFAOYNA-N
reychler's acid, camphorsulfonic acid, d-camphorsulfonic acid, camphersulfosaeure, --10-camphorsulfonic acid, 2-oxobornane-10-sulphonic acid, d-10-camphorsulfonic acid, 7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, l-camphor-10-sulfonic acid, +-10-camphorsulfonic acid
18462
CHEBI:55379
(7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl)methanesulfonic acid
CC1(C2CCC1(C(=O)C2)CS(=O)(=O)O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Absorbance (20% aq. soln.)
Melting Point >195°C
Color Beige
Loss on Drying 1.5% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 100 g
Beilstein 11, 316
Fieser 14,71; 15,68; 16,58
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in glacial acetic acid,slightly soluble in ethyl acetate,practically insoluble in ether
Formula Weight 232.29
Clarity (5% in water) Clear solution
Percent Purity 98%
Physical Form Granular Powder
Chemical Name or Material DL-10-Camphorsulfonic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • DL-10-Camphorsulfonic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'